Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Thin Bar Hexagonal 300 mesh Copper Grid
SDP
Catalog No. 501903217
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper
Catalog No. 50-190-3217 Supplier Electron Microscopy Sciences Supplier No. CFTH300CUUL
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper