Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on Thin Bar Hexagonal 400 mesh Gold Grid
SDP
Catalog No. 501903234
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold
Catalog No. 50-190-3234 Supplier Electron Microscopy Sciences Supplier No. CFTH400AUET
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold