Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Thin Bar Hexagonal 400 mesh Copper Grid
SDP
Catalog No. 501900869
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper
Catalog No. 50-190-0869 Supplier Electron Microscopy Sciences Supplier No. CFTH400CU50
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper