Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Thin Bar Hexagonal 400 mesh Nickel Grid
SDP
Catalog No. 501903229
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel
Catalog No. 50-190-3229 Supplier Electron Microscopy Sciences Supplier No. CFTH400NIUL
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel