Learn More
Electron Microscopy Sciences Carbon Support Film 3-4 nm thick on Thin Bar Hexagonal 600 mesh Gold Grid
SDP
Catalog No. 501903241
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold
Catalog No. 50-190-3241 Supplier Electron Microscopy Sciences Supplier No. CFTH600AUUL
A thin film of pure carbon deposited on one side of the thin bar hexagonal grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold