Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Thin Bar Square 400 mesh Nickel Grid
SDP
Catalog No. 501900846
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel
Catalog No. 50-190-0846 Supplier Electron Microscopy Sciences Supplier No. CFT400NI25
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Nickel