Learn More
Electron Microscopy Sciences Carbon Support Film 5-6 nm thick on Thin Bar Square 1000 mesh Copper Grid
SDP
Catalog No. 501900851
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper
Catalog No. 50-190-0851 Supplier Electron Microscopy Sciences Supplier No. CFT1000CU50
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Standard thickness is 5-6 nm.
Grid: Copper