Learn More
Electron Microscopy Sciences Carbon Support Film 10 nm thick on Thin Bar Square 1000 mesh Nickel Grid
SDP
Catalog No. 501903203
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Nickel
Catalog No. 50-190-3203 Supplier Electron Microscopy Sciences Supplier No. CFT1000NITH
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Nickel