Learn More
Electron Microscopy Sciences Carbon Support Film 20-30 nm thick on Thin Bar Square 400 mesh Gold Grid
SDP
Catalog No. 501903198
Encompass_Preferred
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold
Catalog No. 50-190-3198 Supplier Electron Microscopy Sciences Supplier No. CFT400AUET
A thin film of pure carbon deposited on one side of the thin bar square grid. All of our coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold