Learn More
MSC CRC Citrus Degreaser, Low VOC, NSF C1, Aerosol, 15 oz
Catalog No. 502869809
Encompass
CRC Citrus Degreaser is a new Low VOC Formula product that effectively removes grease, oil, and lubricants without the need for water wash-off. It features d-Limonene and a petroleum blended degreaser, offering controlled evaporation that leaves no residue. It is non-corrosive, non-abrasive, and non-staining, making it suitable for a variety of applications.
  • D-Limonene and petroleum blended degreaser
  • Controlled evaporation, leaves no residue
  • Non-corrosive, non-abrasive, and non-staining
  • NSF C1 registered
  • Pleasant citrus scent
  • Removes grease, oil, and lubricants with no water wash-off
Catalog No. 50-286-9809 Supplier MSC Supplier No. 84275452
CRC Citrus Degreaser is a new Low VOC Formula product that effectively removes grease, oil, and lubricants without the need for water wash-off. It features d-Limonene and a petroleum blended degreaser, offering controlled evaporation that leaves no residue. It is non-corrosive, non-abrasive, and non-staining, making it suitable for a variety of applications.
  • D-Limonene and petroleum blended degreaser
  • Controlled evaporation, leaves no residue
  • Non-corrosive, non-abrasive, and non-staining
  • NSF C1 registered
  • Pleasant citrus scent
  • Removes grease, oil, and lubricants with no water wash-off