Learn More
Grainger CRC Precision Cleaner: Aerosol Spray Can, 13 oz, Liquid, No Series, Hard Nonporous Surfaces
Catalog No. NC3888175
Encompass_Preferred
Contact cleaners, also called precision cleaners, remove dust, dirt, oil, flux residue, and other contaminants from sensitive electronics and electrical components without excessive scrubbing or wiping. These fast-drying cleaners improve conductivity by removing contaminants from the energized surfaces of electronic components, devices, and equipment.
Catalog No. NC3888175 Supplier Grainger Supplier No. 32UN52
Contact cleaners, also called precision cleaners, remove dust, dirt, oil, flux residue, and other contaminants from sensitive electronics and electrical components without excessive scrubbing or wiping. These fast-drying cleaners improve conductivity by removing contaminants from the energized surfaces of electronic components, devices, and equipment.