Learn More
Thermo Scientific Thermostat 1EA
Catalog No. NC1331779 Shop All Thermo Scientific Products
Encompass
A 4mm x 150mm ion chromatography column containing continuously regenerated divinylbenzene cation exchange resin for separating and analyzing monovalent and divalent cations with fast kinetics, high capacity and low baseline noise.
Catalog No. NC1331779 Supplier Thermo Scientific Supplier No. 088663
A 4mm x 150mm ion chromatography column containing continuously regenerated divinylbenzene cation exchange resin for separating and analyzing monovalent and divalent cations with fast kinetics, high capacity and low baseline noise.