Learn More
Thermo Scientific CRD 200 (2mm)
Supplier:  Thermo Scientific 062986
Encompass
Boost sensitivity and improve linearity in your ion chromatography applications by removing carbonate background noise.
Catalog No. NC0363776