Learn More
Electron Microscopy Sciences Cryogenic Cardboard Box and Lid for 15 or 50 ml tubes, 145 x 145 x 122 mm
SDP
Catalog No. 501926810
Encompass_Preferred
Low-cost choice with excellent low-temperature storage properties. One-size box safely stores 15 and 50 ml tubes in LN2 or mechanical freezers.
Catalog No. 50-192-6810 Supplier Electron Microscopy Sciences Supplier No. 41004
Low-cost choice with excellent low-temperature storage properties. One-size box safely stores 15 and 50 ml tubes in LN2 or mechanical freezers.