Learn More
Electron Microscopy Sciences Cardboard Freezer Box 2" with 81 well divider
SDP
Catalog No. 501927419
Encompass_Preferred
For use with Stainless Steel Freezer Racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Cardboard boxes can be labelled with Cryo-Tags for easy identification.
Catalog No. 50-192-7419 Supplier Electron Microscopy Sciences Supplier No. 6161595
For use with Stainless Steel Freezer Racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Cardboard boxes can be labelled with Cryo-Tags for easy identification.