Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Cardboard Well Slide Double Depth with Paper Top, 1 Well 15 mm, White
SDP

Supplier:  Electron Microscopy Sciences 6351514

Encompass_Preferred

EMS introduces these unique cardboard "well" slides. Complete with glass covered slide and aluminum holder, these slides are less prone to static as compared to similar plastic slides.
Wells: 1 Well
Size: 15 mm
Color: White
Slide: 74.7 mm x 25.3 mm x 3.6 mm (H)

Catalog No. 50-193-0108


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.