Learn More
MSC Atrix Cordless Backpack Vacuum, 26V, 8 qt, HEPA, 1 ea
Catalog No. 502817986
Encompass
This cordless backpack vacuum is designed for janitorial, sanitation, contract cleaning, offices, and warehouses, especially in locations without an AC power source or where cords could be a safety hazard.
  • Low-profile design with padded harness, accessory holders, and adjustable chest strap for comfort
  • Operates on 26V with 300 watts cleaning power, producing 70 CFM
  • Provides 60 minutes of run time and a 4-hour quick charge
  • Features four levels of filtration, including an eight-quart HEPA filtration bag
  • Includes a 1.5 in industry standard hose and various cleaning utensils
Catalog No. 50-281-7986 Supplier MSC Supplier No. 14406805
This cordless backpack vacuum is designed for janitorial, sanitation, contract cleaning, offices, and warehouses, especially in locations without an AC power source or where cords could be a safety hazard.
  • Low-profile design with padded harness, accessory holders, and adjustable chest strap for comfort
  • Operates on 26V with 300 watts cleaning power, producing 70 CFM
  • Provides 60 minutes of run time and a 4-hour quick charge
  • Features four levels of filtration, including an eight-quart HEPA filtration bag
  • Includes a 1.5 in industry standard hose and various cleaning utensils