Learn More
MSC DeWALT Wet/Dry Vacuum, Battery-Powered, 0.1 gal, HEPA Filter, 1 ea
Catalog No. 502866825
Encompass
This wet/dry vacuum is battery-powered, offering a 0.1 gal collection capacity and HEPA filtration within a durable plastic tank. Designed for versatility, it is suitable for various clean-up tasks.
  • Battery-powered operation
  • HEPA filtration for fine dust collection
  • Collection capacity of 0.1 gal
  • Dust cup vacuum collection type
  • Plastic tank material
  • Maximum air flow of 46 CFM
  • Hose diameter of 1.2 in
  • Includes floor nozzle, extension tube, crevice nozzle, accessory bag, round brush, flexible hose, and gulper brush
Catalog No. 50-286-6825 Supplier MSC Supplier No. 16299133
This wet/dry vacuum is battery-powered, offering a 0.1 gal collection capacity and HEPA filtration within a durable plastic tank. Designed for versatility, it is suitable for various clean-up tasks.
  • Battery-powered operation
  • HEPA filtration for fine dust collection
  • Collection capacity of 0.1 gal
  • Dust cup vacuum collection type
  • Plastic tank material
  • Maximum air flow of 46 CFM
  • Hose diameter of 1.2 in
  • Includes floor nozzle, extension tube, crevice nozzle, accessory bag, round brush, flexible hose, and gulper brush