Learn More
MSC DeWALT Cordless Dry Hand Vacuum, 20V, HEPA Filtration, Tool Only
Catalog No. 502838476
Encompass
The 20V cordless dry hand vacuum is a powerful jobsite cleaning solution with 46 CFM, designed to pick up common debris. It includes 6 versatile cleaning attachments and a HEPA filter for efficient fine dust cleanup (OSHA Table 1 compliant). Features include an LED light and a belt hook for hands-free operation. Battery and charger sold separately.
  • 46 CFM provides higher suction for efficient clean-up.
  • 6 accessory cleaning solutions included.
  • Drawstring accessory bag for organized storage.
  • Easy empty canister minimizes dust exposure.
  • HEPA filtration for clean fine dust cleanup.
  • Bright LED light illuminates dark areas.
  • Belt hook for hands-free operation.
Catalog No. 50-283-8476 Supplier MSC Supplier No. 67964189
The 20V cordless dry hand vacuum is a powerful jobsite cleaning solution with 46 CFM, designed to pick up common debris. It includes 6 versatile cleaning attachments and a HEPA filter for efficient fine dust cleanup (OSHA Table 1 compliant). Features include an LED light and a belt hook for hands-free operation. Battery and charger sold separately.
  • 46 CFM provides higher suction for efficient clean-up.
  • 6 accessory cleaning solutions included.
  • Drawstring accessory bag for organized storage.
  • Easy empty canister minimizes dust exposure.
  • HEPA filtration for clean fine dust cleanup.
  • Bright LED light illuminates dark areas.
  • Belt hook for hands-free operation.