Learn More
Cayman Chemical Cre mRNACap-1m1 1mg
SDP
Catalog No. 502945615
Encompass_Preferred
An mRNA for the expression of Cre; encapsulation in LNPs can be used for mRNA delivery and expression of Cre in vitro or in vivo
Catalog No. 50-294-5615 Supplier Cayman Chemical Supplier No. 419631 MG
An mRNA for the expression of Cre; encapsulation in LNPs can be used for mRNA delivery and expression of Cre in vitro or in vivo