Learn More
Cayman Chemical Cre mRNACap-1UTP 100ug
SDP
Catalog No. 502943975
Encompass_Preferred
An mRNA for the expression of Cre; encapsulation in LNPs can be used for mRNA delivery and expression of Cre in vitro or in vivo
Catalog No. 50-294-3975 Supplier Cayman Chemical Supplier No. 39802100 UG
An mRNA for the expression of Cre; encapsulation in LNPs can be used for mRNA delivery and expression of Cre in vitro or in vivo