Learn More
Cayman Chemical Cre mRNACap-1UTP 1mg
SDP
Catalog No. 502944927
Encompass_Preferred
An mRNA for the expression of Cre; encapsulation in LNPs can be used for mRNA delivery and expression of Cre in vitro or in vivo
Catalog No. 50-294-4927 Supplier Cayman Chemical Supplier No. 398021 MG
An mRNA for the expression of Cre; encapsulation in LNPs can be used for mRNA delivery and expression of Cre in vitro or in vivo