Learn More
Takara Bio Cre Recombinase Gesicles | 200 uL
SDP
Supplier:  Takara Bio 631449
Encompass
Cre Recombinase Gesicles are cell-derived nanovesicles used to deliver active Cre recombinase protein directly into target cells. When they are applied to your target cells, the gesicles fuse with the plasma membrane and deliver a temporary dose of Cre recombinase, which facilitates recombination at loxP sites contained within the cell. As an added benefit, Cre Recombinase Gesicles contain CherryPicker protein, a membrane-targeted version of the mCherry fluorescent protein that helps to confirm the delivery of the Cre recombinase to your target cells.
Catalog No. NC1442408