Learn More
Enzo Life Sciences Cre-recombinase polyclonal antibody (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2040200
Encompass_Preferred
Immunogen: Recombinant full length cre-recombinase. Specificity: Recognizes cre-recombinase. Applications: IHC, WB. Purity: Protein A purified. Formulation: Liquid. In PBS containing 1% BSA and 0.1% sodium azide.
Catalog No. 50-201-2754