Learn More
ACROBiosystems Recombinant Protein;Human C-Reactive Protein (CRP) Protein (HPLC-verified);HEK293;1MG;CRP,CRP,C-reactive Protein
Supplier:  ACROBiosystems CRPH52261MG
Encompass_Preferred
Recombinant Protein;Human C-Reactive Protein (CRP) Protein (HPLC-verified);1MG;Human CRP, His Tag (CRP-H5226) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Pro 224 (Accession # NP_000558.2).;This protein carries a polyhistidine tag at the C-terminus, and has a calculated MW of 24 kDa. The predicted N-terminus is Phe 17. The reducing (R) protein migrates as 26 kDa in SDS-PAGE. More than 90% rhCRP exists as active homopentamer in analytical SEC-HPLC assay.
Catalog No. 50-165-7527