Learn More
Med Vet International Skin Protectant 3M Cavilon 2 Gram Individual Packet Unscented Cream Chg Compatible, 3353, 240/Case
Catalog No. 502831945
Encompass_Preferred
The moisturizing barrier cream contains dimethicone to keep skin protected This incontinence barrier cream supplies the skin with moisture to help keep it healthy and free of incontinence-associated dermatitis It disappears into the user's skin so it won't prevent tapes and dressings from adhering 3M™ Cavilon™ durable barrier cream is fragrance free and hypoallergenic to prevent skin irritation It's CHG-compatible for use with germicidal soaps and cleansers It resists washing off so that its protection will last
Catalog No. 50-283-1945 Supplier Med Vet International Supplier No. 798691CS
The moisturizing barrier cream contains dimethicone to keep skin protected This incontinence barrier cream supplies the skin with moisture to help keep it healthy and free of incontinence-associated dermatitis It disappears into the user's skin so it won't prevent tapes and dressings from adhering 3M™ Cavilon™ durable barrier cream is fragrance free and hypoallergenic to prevent skin irritation It's CHG-compatible for use with germicidal soaps and cleansers It resists washing off so that its protection will last