Learn More
Med Vet International Skin Protectant 3M Cavilon 3.25oz Tube Unscented Cream Chg Compatible, 3355, Each
Catalog No. 502829370
Encompass_Preferred
The 3M™ Cavilon™ skin protectant defends vulnerable perineal skin against erosion It doesn't leave behind a residue, which allows adhesives to adhere It contains dimethicone, which conditions the skin It is CHG compatible and won't interfere with antiseptics The unscented cream lasts a long time, requiring fewer applications The Cavilon™ barrier cream is hypoallergenic It comes in a 3.25 ounce tube for convenience
Catalog No. 50-282-9370 Supplier Med Vet International Supplier No. 806588EA
The 3M™ Cavilon™ skin protectant defends vulnerable perineal skin against erosion It doesn't leave behind a residue, which allows adhesives to adhere It contains dimethicone, which conditions the skin It is CHG compatible and won't interfere with antiseptics The unscented cream lasts a long time, requiring fewer applications The Cavilon™ barrier cream is hypoallergenic It comes in a 3.25 ounce tube for convenience