Learn More
Med Vet International Skin Protectant 3M Cavilon 1oz Tube Unscented Cream Chg Compatible, 3354, 48/Case
Catalog No. 502829919
Encompass_Preferred
The 28-gram moisturizing barrier cream contains dimethicone to keep skin protected This incontinence barrier cream supplies the skin with moisture to help keep it healthy and free of incontinence-associated dermatitis It disappears into the user's skin so it won't prevent tapes and dressings from adhering This fragrance-free 3M™ Cavilon™ durable barrier cream hypoallergenic to prevent skin irritation It's CHG-compatible for use with germicidal soaps and cleansers It resists washing off so that its protection will last
Catalog No. 50-282-9919 Supplier Med Vet International Supplier No. 842937CS
The 28-gram moisturizing barrier cream contains dimethicone to keep skin protected This incontinence barrier cream supplies the skin with moisture to help keep it healthy and free of incontinence-associated dermatitis It disappears into the user's skin so it won't prevent tapes and dressings from adhering This fragrance-free 3M™ Cavilon™ durable barrier cream hypoallergenic to prevent skin irritation It's CHG-compatible for use with germicidal soaps and cleansers It resists washing off so that its protection will last