Learn More
Cayman Chemical Creatine Phosphate (sodium salt hydrate), Purity: ≥95%, CAS: 19333-65-4, 10 g
SDP
Catalog No. 502916691
Encompass_Preferred
A phosphorylated form of creatine; reduces hydrogen peroxide- and ferrous ion-induced increases in MDA levels in rat heart microsomes at 10 mM; prevents hydrogen peroxide-induced decreases in aortic flow, minute work, and peak aortic pressure in isolated perfused rat hearts at 10 mM; brain levels decrease following mild traumatic brain injury in rats
Catalog No. 50-291-6691 Supplier Cayman Chemical Supplier No. 3780310 G
A phosphorylated form of creatine; reduces hydrogen peroxide- and ferrous ion-induced increases in MDA levels in rat heart microsomes at 10 mM; prevents hydrogen peroxide-induced decreases in aortic flow, minute work, and peak aortic pressure in isolated perfused rat hearts at 10 mM; brain levels decrease following mild traumatic brain injury in rats