Learn More
Medchemexpress LLC Creatine-13C | 687996-99-2 | 98.5% | 132.13 | 1 MG
SDP
Catalog No. 5000367042
Encompass_Preferred
Creatine-13C is the 13C labeled Creatine, an endogenous amino acid derivative that plays an important role in cellular energy, especially in muscle and brain. This product has been tested and complies with the given specifications.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-67042 Supplier Medchemexpress LLC Supplier No. HYW010388S1MG
Creatine-13C is the 13C labeled Creatine, an endogenous amino acid derivative that plays an important role in cellular energy, especially in muscle and brain. This product has been tested and complies with the given specifications.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS