Learn More
Medchemexpress LLC Creatine-13C | 687996-99-2 | 98.5% | 132.13 | 25 MG
SDP
Catalog No. 5000451485
Encompass_Preferred
Creatine-13C is a 13C labeled Creatine, which is an endogenous amino acid derivative. It plays a significant role in cellular energy, particularly in muscle and brain. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotope of carbon (13C) incorporated into the molecule.
Catalog No. 50-004-51485 Supplier Medchemexpress LLC Supplier No. 25MGHYW010388S
Creatine-13C is a 13C labeled Creatine, which is an endogenous amino acid derivative. It plays a significant role in cellular energy, particularly in muscle and brain. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotope of carbon (13C) incorporated into the molecule.