Learn More
Medchemexpress LLC Creatine-d3 | 143827-19-4 | 99.5% | 134.15 | 10 MG
SDP
Catalog No. 5000367043
Encompass_Preferred
Creatine-d3 is the deuterium labeled Creatine (HY-W010388). Creatine, an endogenous amino acid derivative, plays an important role in cellular energy, especially in muscle and brain. For research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-67043 Supplier Medchemexpress LLC Supplier No. HYW010388S110MG
Creatine-d3 is the deuterium labeled Creatine (HY-W010388). Creatine, an endogenous amino acid derivative, plays an important role in cellular energy, especially in muscle and brain. For research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.