Learn More
Medchemexpress LLC Creatine-d3 | 143827-19-4 | 99.5% | 134.15 | 25 MG
SDP
Catalog No. 5000366712
Encompass_Preferred
Creatine-d3 is a deuterium-labeled Creatine. Creatine, an endogenous amino acid derivative, plays an important role in cellular energy, particularly in muscle and brain.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66712 Supplier Medchemexpress LLC Supplier No. 25MGHYW010388S1
Creatine-d3 is a deuterium-labeled Creatine. Creatine, an endogenous amino acid derivative, plays an important role in cellular energy, particularly in muscle and brain.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.