Learn More
TARGETMOL CHEMICALS INC Creatine-d3 hydrate 1MG
SDP
Catalog No. 502879074
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Creatine-d3 hydrate (CREATINE-(METHYL-D3) MONOHYDRATE) is a deuterium labeled Creatine hydrate. Under physiological and in pathological conditions, Creatine hydrate is pivotal in energy metabolism of muscle and brain cells. Purity 92.96%
Catalog No. 50-287-9074 Supplier TARGETMOL CHEMICALS INC Supplier No. T90871MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Creatine-d3 hydrate (CREATINE-(METHYL-D3) MONOHYDRATE) is a deuterium labeled Creatine hydrate. Under physiological and in pathological conditions, Creatine hydrate is pivotal in energy metabolism of muscle and brain cells. Purity 92.96%