Learn More
Medchemexpress LLC Creatine-d3 hydrate | 284664-86-4 | 99.0% | 152.17 | 10 MG
SDP
Catalog No. 5000453895
Encompass_Preferred
Creatine-d3 (hydrate) is a deuterium labeled Creatine hydrate. Creatine hydrate is pivotal in energy metabolism of muscle and brain cells, both in physiological and in pathological conditions. It is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53895 Supplier Medchemexpress LLC Supplier No. HYW010388AS10MG
Creatine-d3 (hydrate) is a deuterium labeled Creatine hydrate. Creatine hydrate is pivotal in energy metabolism of muscle and brain cells, both in physiological and in pathological conditions. It is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.