Learn More
Medchemexpress LLC Creatine-d5 monohydrate | 99.9% | 154.18 g/mol | C4H6D5N3O3 | 1 MG
SDP
Catalog No. 5000300410
Encompass_Preferred
Creatine-d5 monohydrate is the deuterium-labeled analog of creatine monohydrate provided as a stable-isotope reagent for research use. It serves as an internal standard for mass spectrometry, tracer experiments, and metabolic studies where isotopic labeling enables precise quantitation and pathway tracking. Supplied as a powder in small analytical pack sizes with documented purity and storage recommendations.
Catalog No. 50-003-00410 Supplier Medchemexpress LLC Supplier No. HYW017462S1MG
Creatine-d5 monohydrate is the deuterium-labeled analog of creatine monohydrate provided as a stable-isotope reagent for research use. It serves as an internal standard for mass spectrometry, tracer experiments, and metabolic studies where isotopic labeling enables precise quantitation and pathway tracking. Supplied as a powder in small analytical pack sizes with documented purity and storage recommendations.