Learn More
Medchemexpress LLC Creatine-d5 monohydrate | 99.9% | 154.18 g/mol | C4H6D5N3O3 | 5 MG
SDP
Catalog No. 5000300607
Encompass_Preferred
Creatine-d5 monohydrate is a deuterium-labeled form of creatine monohydrate used as a stable-isotope internal standard and analytical reference for quantitative LC-MS/MS and tracer studies. It separates from unlabeled creatine, enabling accurate quantitation in biological, food, and analytical matrices.
  • Stable-isotope labeled for use as an internal standard.
  • High purity (99.93%) for reliable analytical performance.
  • Suitable for LC-MS/MS quantitation and tracer studies.
  • Small package sizes support method development and validation.
  • Provides a clear mass shift from unlabeled analyte for peak identification.
Catalog No. 50-003-00607 Supplier Medchemexpress LLC Supplier No. HYW017462S5MG
Creatine-d5 monohydrate is a deuterium-labeled form of creatine monohydrate used as a stable-isotope internal standard and analytical reference for quantitative LC-MS/MS and tracer studies. It separates from unlabeled creatine, enabling accurate quantitation in biological, food, and analytical matrices.
  • Stable-isotope labeled for use as an internal standard.
  • High purity (99.93%) for reliable analytical performance.
  • Suitable for LC-MS/MS quantitation and tracer studies.
  • Small package sizes support method development and validation.
  • Provides a clear mass shift from unlabeled analyte for peak identification.