Learn More
Sigma Aldrich Fine Chemicals Biosciences Creatine Kinase (CK), 100 mg
Catalog No. NC1366773
Encompass
Creatine kinase has been used as a component of translation buffer used in in vitro translation assay and as a component of energy regeneration mix.
Catalog No. NC1366773 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127566001
Creatine kinase has been used as a component of translation buffer used in in vitro translation assay and as a component of energy regeneration mix.