Learn More
Sigma Aldrich Fine Chemicals Biosciences Creatine Kinase Activity Assay Kit
Catalog No. NC1636581
Encompass
Sufficient for 100 tests (Ultraviolet), Synonym: CK Activity Assay Kit, Creatine Phosphokinase Activity Assay Kit; Creatine Kinase (CK), also known as phosphocreatine kinase, is an enzyme that catalyzes the transfer of one phosphate group from ATP to creatine generating phosphocreatine, an important energy reservoir in muscle and brain tissue.
Catalog No. NC1636581 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1161KT
Sufficient for 100 tests (Ultraviolet), Synonym: CK Activity Assay Kit, Creatine Phosphokinase Activity Assay Kit; Creatine Kinase (CK), also known as phosphocreatine kinase, is an enzyme that catalyzes the transfer of one phosphate group from ATP to creatine generating phosphocreatine, an important energy reservoir in muscle and brain tissue.