Learn More
Sigma Aldrich Fine Chemicals Biosciences Creatine Kinase CK from ra
Catalog No. 502907874
Encompass
ATPcreatine N-phosphotransferaseCreatine kinase (CK) has a molar mass of 85kDa. It exists as a dimer in the cytosolic form and as a dimer and an octamer in the mitochondrial form. The cytosolic forms are named as MM BB and MB where M and B correspond to muscle and brain respectively. CK from rabbit muscle is a dimer containing two identical polypeptide chains of 380 amino acids. Each subunit has four cysteines Cys73 Cys145 Cys253 and Cys282 but lacks inter- and intra-chain disulfide bonds.
Catalog No. 50-290-7874 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10736988001
ATPcreatine N-phosphotransferaseCreatine kinase (CK) has a molar mass of 85kDa. It exists as a dimer in the cytosolic form and as a dimer and an octamer in the mitochondrial form. The cytosolic forms are named as MM BB and MB where M and B correspond to muscle and brain respectively. CK from rabbit muscle is a dimer containing two identical polypeptide chains of 380 amino acids. Each subunit has four cysteines Cys73 Cys145 Cys253 and Cys282 but lacks inter- and intra-chain disulfide bonds.