Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Creatine kinase M-type (mouse), recombinant, His-tagged | >90.0% | 20UG
SDP

Catalog No. 5000177019
Encompass_Preferred

Recombinant mouse creatine kinase M-type (CKM) is expressed in E. coli and purified with an N-terminal His tag; it is supplied lyophilized for research use and formulated for stability. The product has >90% purity and an approximate molecular weight of 49.0 kDa; store frozen and follow reconstitution instructions for enzymatic or immunological applications.

  • Recombinant mouse CKM with N-terminal His tag.
  • High purity (>90%) determined by reducing SDS-PAGE.
  • Molecular weight approximately 49.0 kDa.
  • Supplied lyophilized for convenient storage and shipping.
  • Formulated in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Suitable for enzymatic assays, antibody production, and biochemical studies.
  • Store at -20°C for long-term stability; aliquot to avoid freeze-thaw cycles.

Catalog No. 50-001-77019 Supplier Medchemexpress LLC Supplier No. HYP70057720UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse creatine kinase M-type (CKM) is expressed in E. coli and purified with an N-terminal His tag; it is supplied lyophilized for research use and formulated for stability. The product has >90% purity and an approximate molecular weight of 49.0 kDa; store frozen and follow reconstitution instructions for enzymatic or immunological applications.

  • Recombinant mouse CKM with N-terminal His tag.
  • High purity (>90%) determined by reducing SDS-PAGE.
  • Molecular weight approximately 49.0 kDa.
  • Supplied lyophilized for convenient storage and shipping.
  • Formulated in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Suitable for enzymatic assays, antibody production, and biochemical studies.
  • Store at -20°C for long-term stability; aliquot to avoid freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.