Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Creatine kinase M-type/CKM protein, mouse (His) | >90.0% | 50UG
SDP

Catalog No. 5000174964
Encompass_Preferred

Recombinant mouse creatine kinase M-type (CKM) is supplied as a lyophilized, His-tagged protein expressed in E. coli for research use. CKM catalyzes reversible phosphate transfer between ATP and creatine phosphate and supports energy buffering in high-demand tissues. The material is provided with high purity and low endotoxin and is intended for biochemical assays, enzymology, and antibody validation. Store frozen and follow supplier reconstitution recommendations.

  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • Expressed in E. coli with an N-terminal 6xHis tag to facilitate purification.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Molecular weight approximately 49.0 kDa, consistent with the CKM isoform.
  • Recommended storage at -20°C; stable short term at 4°C after reconstitution.

Catalog No. 50-001-74964 Supplier Medchemexpress LLC Supplier No. HYP70057750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse creatine kinase M-type (CKM) is supplied as a lyophilized, His-tagged protein expressed in E. coli for research use. CKM catalyzes reversible phosphate transfer between ATP and creatine phosphate and supports energy buffering in high-demand tissues. The material is provided with high purity and low endotoxin and is intended for biochemical assays, enzymology, and antibody validation. Store frozen and follow supplier reconstitution recommendations.

  • High purity (>90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • Expressed in E. coli with an N-terminal 6xHis tag to facilitate purification.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Molecular weight approximately 49.0 kDa, consistent with the CKM isoform.
  • Recommended storage at -20°C; stable short term at 4°C after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.