Learn More
Cusabio Technology LLC creatine kinase MB isoenzyme
Catalog No. 5000050294
Encompass_Preferred
This kit detects CK-MB in pig serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 2 35 mU/mL to 150 mU/mL with a sensitivity of 0 58 mU/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Results are determined by absorbance at 450 nm ensuring reliable and specific quantification of CK-MB levels for applications in clinical diagnostics or research studies involving myocardial injury assessment The kit is designed for straightforward implementation in laboratory settings while maintaining high reproducibility across biological matrices
Catalog No. 50-000-50294 Supplier Cusabio Technology LLC Supplier No. CSBEQ027653PI
This kit detects CK-MB in pig serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 2 35 mU/mL to 150 mU/mL with a sensitivity of 0 58 mU/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Results are determined by absorbance at 450 nm ensuring reliable and specific quantification of CK-MB levels for applications in clinical diagnostics or research studies involving myocardial injury assessment The kit is designed for straightforward implementation in laboratory settings while maintaining high reproducibility across biological matrices