Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Creatine Kinase-MB Kit, Calibrator Grade Standard, Recombinant Protein

Catalog No. piakc0325
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIAKC0325 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIAKC0325 Supplier Invitrogen™ Supplier No. AKC0325

Recombinant Protein

NULL

The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis and is an important serum marker for myocardial infarction. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in striated muscle as well as in other tissues, and as a heterodimer with a similar brain isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. Kit includes 100ug of lyophilized recombinant Human CK-MB protein and 3 bottles of diluting buffer (1 mL each). Detailed protocol is included with CofA, all reagents are included. The protocol describes the rCK-MB hydration process and dilution steps required for the determination of rCK-MB value in a test CK-MB immunoassay. The results should be used to assign CK-MB values of the calibrators for the test immunoassay. Quality Specifications: Purity is greater than or equal to 95% by SDS PAGE. Specific activity is greater than 950 U/mg by Genzyme Manual Spectrophotometer. Storage: Diluent should be stored at 4°C. Lyophilized protein should be stored at 4°C short-term, and long-term at -20°C. Reconstitution: reconstitute 100ug of Recombinant Human CK-MB, Calibrator Grade with 1 mL of CK-MB Diluent. Mix gently, and allow to stand for at least 15 minutes prior to use. After using, replace stopper and store at 2-8°C to achieve at least3 weeks of reconstituted stability.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control
Formulation protein
Name Creatine Kinase-MB Kit, Calibrator Grade Standard
Quantity 100 μg
Source E. Coli
Storage Requirements Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Regulatory Status RUO
Gene Alias Creatine kinase, M chain; M-CK; CKMM
Common Name Creatine Kinase-MB Kit, Calibrator Grade Standard
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.