Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Creatine kinase MB (rCK-MB), recombinant human (CK-MB) | 9001-15-4 | 100 UG
SDP

Catalog No. 5000399899
Encompass_Preferred

Creatine Kinase MB (rCK-MB), Recombinant Human (CK-MB) is a biological material or organic compound that can be used in life science research. This product is intended for research use only.

  • Appearance: liquid, colorless to light yellow.
  • Enzyme activity: > 400 U/mL.
  • Unit definition: one unit is defined as the amount of enzyme that catalyzes the transphosphorylation of one micromole of phosphate from creatine phosphate to ADP per minute at 37°C.
  • Shipping with dry ice.
  • Store under recommended conditions in the certificate of analysis.

Catalog No. 50-003-99899 Supplier Medchemexpress LLC Supplier No. HYP2799D100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creatine Kinase MB (rCK-MB), Recombinant Human (CK-MB) is a biological material or organic compound that can be used in life science research. This product is intended for research use only.

  • Appearance: liquid, colorless to light yellow.
  • Enzyme activity: > 400 U/mL.
  • Unit definition: one unit is defined as the amount of enzyme that catalyzes the transphosphorylation of one micromole of phosphate from creatine phosphate to ADP per minute at 37°C.
  • Shipping with dry ice.
  • Store under recommended conditions in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.