Learn More
Cambridge Isotope Laboratories Creatine (Methyl-D3) | 143827-20-7 | MFCD00190410 | 0.25 g
Catalog No. NC1529463
Encompass
Creatine (Methyl-D3) | Purity: 97% | Mol Wt: 134.15 | 143827-20-7 | MFCD00190410 | 0.25 g
Catalog No. NC1529463 Supplier Cambridge Isotope Laboratories Supplier No. DLM13020.25
Creatine (Methyl-D3) | Purity: 97% | Mol Wt: 134.15 | 143827-20-7 | MFCD00190410 | 0.25 g