Learn More
Apexbio Technology LLC Creatine monohydrate 6020-87-7 100mg
SDP
Catalog No. 502240693
Encompass_Preferred
Creatine monohydrate (CAS 6020-87-7) is an endogenous amino acid derivative that modulates cellular energy metabolism primarily through its involvement in the phosphocreatine pathway It facilitates the rapid regeneration of adenosine triphosphate (ATP) by serving as a phosphate donor in muscle and neural tissues thereby supporting cellular energy demands during periods of increased metabolic activity Due to its capacity to enhance ATP synthesis and influence muscle contractility creatine monohydrate is widely investigated in studies of muscle disorders and neurodegenerative diseases offering a valuable tool for exploring cellular energy homeostasis in various biomedical research contexts
This item is not returnable. View return policy
Catalog No. 50-224-0693 Supplier Apexbio Technology LLC Supplier No. C6421100
Creatine monohydrate (CAS 6020-87-7) is an endogenous amino acid derivative that modulates cellular energy metabolism primarily through its involvement in the phosphocreatine pathway It facilitates the rapid regeneration of adenosine triphosphate (ATP) by serving as a phosphate donor in muscle and neural tissues thereby supporting cellular energy demands during periods of increased metabolic activity Due to its capacity to enhance ATP synthesis and influence muscle contractility creatine monohydrate is widely investigated in studies of muscle disorders and neurodegenerative diseases offering a valuable tool for exploring cellular energy homeostasis in various biomedical research contexts