Learn More
Abcam Creatine phosphate disodium salt tetrahydrate, Phosphate reservoir, 5G
Catalog No. 502704793
Encompass_Preferred
MW 327.14 g/mol, Purity >99%. Phosphate reservoir; used to regenerate ATP during skeletal muscle contraction.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4793 Supplier Abcam Supplier No. AB1462555G
MW 327.14 g/mol, Purity >99%. Phosphate reservoir; used to regenerate ATP during skeletal muscle contraction.
The product is subject to the following: Abcam Restricted Use Statement