Learn More
Abcam Creatine phosphate disodium salt tetrahydrate, Phosphate reservoir, 25G
Catalog No. 502704794
Encompass_Preferred
MW 327.14 g/mol, Purity >99%. Phosphate reservoir; used to regenerate ATP during skeletal muscle contraction.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4794 Supplier Abcam Supplier No. AB14625525G
MW 327.14 g/mol, Purity >99%. Phosphate reservoir; used to regenerate ATP during skeletal muscle contraction.
The product is subject to the following: Abcam Restricted Use Statement