Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine phosphate, disodium salt, 99.12%, MP Biomedicals™

Catalog No. ICN10052001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10052001 1 g
ICN10052025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10052001 Supplier MP Biomedicals, Inc Supplier No. 0210052001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 922-32-7
Molecular Formula C4H8N3Na2O5P
Molecular Weight (g/mol) 255.08
MDL Number MFCD00150192
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
Synonym creatine phosphate sodium salt
PubChem CID 131887655
IUPAC Name disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 98-100%
Formula Weight 327.2
Chemical Name or Material Creatine phosphate, disodium salt
Recommended Storage Store at - 0°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.